missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pentene, cis + trans, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
Specifications
Density | 0.65 |
Boiling Point | 36°C to 38°C |
Flash Point | −45°C (−49°F) |
Odor | Petroleum-like |
Linear Formula | CH3CH2CH=CHCH3 |
Refractive Index | 1.38 |
Quantity | 25 g |
UN Number | UN3295 |
Beilstein | 969151 |
Merck Index | 14,7123 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'