missing translation for 'onlineSavingsMsg'
Learn More

2-Pentene, cis + trans, 98%

Product Code. 11463328
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463328 100 g 100g
11453328 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463328 Supplier Thermo Scientific Alfa Aesar Supplier No. A17736.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-68-2 | C5H10 | 70.14 g/mol

2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-68-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009384
InChI Key QMMOXUPEWRXHJS-HYXAFXHYSA-N
Synonym trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
PubChem CID 5326161
SMILES CC\C=C/C

Specifications

Density 0.65
Boiling Point 36°C to 38°C
Flash Point −45°C (−49°F)
Odor Petroleum-like
Linear Formula CH3CH2CH=CHCH3
Refractive Index 1.38
Quantity 100 g
UN Number UN3295
Beilstein 969151
Merck Index 14,7123
Solubility Information Not miscible or difficult to mix in water.
IUPAC Name (E)-pent-2-ene
Formula Weight 70.14
Percent Purity 98%
Chemical Name or Material 2-Pentene, cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.