missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanone, 99+%

Product Code. 11338006
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338006 100 g 100g
11328006 25 g 25g
11348006 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338006 Supplier Thermo Scientific Alfa Aesar Supplier No. L13262.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 107-87-9 | C5H10O | 86.134 g/mol

2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.

Solubility
Miscible with water.

Notes
Incompatible with strong bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-87-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00009400
InChI Key XNLICIUVMPYHGG-UHFFFAOYSA-N
Synonym 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone
PubChem CID 7895
ChEBI CHEBI:16472
IUPAC Name pentan-2-one
SMILES CCCC(=O)C

Specifications

Melting Point -78°C
Density 0.813
Boiling Point 100°C to 101°C
Flash Point 7°C (44°F)
Refractive Index 1.3905
Quantity 100 g
UN Number UN1249
Beilstein 506058
Merck Index 14,6114
Solubility Information Miscible with water.
Formula Weight 86.13
Percent Purity ≥99%
Chemical Name or Material 2-Pentanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.