missing translation for 'onlineSavingsMsg'
Learn More

2-(P-Sulfophenylazo)-1,8-dihydroxy-3,6-naphthalenesulfonic acid trisodium salt, MP Biomedicals™

Product Code. 11445151 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445151 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11445151 Supplier MP Biomedicals Supplier No. 0215672910
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent used for analysis of fluoride, zirconium and thorium

Specifications

CAS 23647-14-5
Color Maroon
Molecular Formula C16H10N2Na3O11S3+
MDL Number MFCD00003952
Quantity 10 g
Synonym spadns, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-4-sulfophenyl azo-, trisodium salt, trisodium 4,5-dihydroxy-3-e-2-4-sulfonatophenyl diazen-1-yl naphthalene-2,7-disulfonate, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-2-4-sulfophenyl diazenyl-, sodium salt 1:3, 2-p-sulfophenylazo-1,8-dihydroxy-3,6-naph-thalenedisulfonic acid
InChI Key LSAWBVPBZOEGOT-UHFFFAOYSA-L
SMILES C1=CC(=CC=C1NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O.[Na+].[Na+].[Na+]
IUPAC Name trisodium;5-hydroxy-4-oxo-3-[(4-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
Molecular Weight (g/mol) 571.408
PubChem CID 73658865
Formula Weight 570.41
Physical Form Powder
Chemical Name or Material 2-(P-Sulfophenylazo)-1, 8-dihydroxy-3, 6-naphthalenesulfonic acid trisodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.