missing translation for 'onlineSavingsMsg'
Learn More

2-Oxovaleric acid, MedChemExpress

Product Code. 16497467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497467 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16497467 Supplier Medchem Express Supplier No. HY113098
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Oxovaleric acid is a keto acid that is found in human blood.

2-Oxovaleric acid is a keto acid that is found in human blood.

Specifications

Chemical Name or Material 2-Oxovaleric acid
Physical Form Liquid
CAS 1821-02-9
Color Colorless
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C5H8O3
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 100 mg/mL (861.18 mM; Need ultrasonic)
SMILES CCCC(C(O)=O)=O
Molecular Weight (g/mol) 116.12
Formula Weight 116.12
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.