missing translation for 'onlineSavingsMsg'
Learn More

2-Oxoindoline-5-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10367584
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367584 250 mg Amber Glass Bottle 250mg
10498593 1 g Amber Glass Bottle 1g
10205664 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367584 Supplier Thermo Scientific Maybridge Supplier No. CC53303CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 199328-31-9
Molecular Formula C8H6ClNO3S
Molecular Weight (g/mol) 231.65
MDL Number MFCD06655924
InChI Key FYBNVKMJOOYPGI-UHFFFAOYSA-N
Synonym 2-oxoindoline-5-sulfonyl chloride, 2-oxo-2,3-dihydro-1h-indole-5-sulfonyl chloride, 2-oxoindoline-5-sulfonylchloride, 2-oxoindoline-5-sulphonyl chloride, 2,3-dihydro-2-oxo-1h-indole-5-sulphonyl chloride, 1h-indole-5-sulfonyl chloride, 2,3-dihydro-2-oxo, 5-chlorosulfonyl indolin-2-one, 5-chlorosulphonyloxindole, 5-chlorosulfonyl oxindole, 5-chlorosufonyl-2-oxindole
PubChem CID 4962506
SMILES ClS(=O)(=O)C1=CC2=C(NC(=O)C2)C=C1

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2-oxo-1,3-dihydroindole-5-sulfonyl chloride
Formula Weight 231.66
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Oxoindoline-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.