missing translation for 'onlineSavingsMsg'
Learn More

2-Oxocyclopentylacetic acid, 97%

Product Code. 11451600
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451600 5 g 5g
11441600 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451600 Supplier Thermo Scientific Alfa Aesar Supplier No. H26977.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1460-38-4 | C7H10O3 | 142.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1460-38-4
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD02093672
InChI Key OLLLIBGOZUPLOK-UHFFFAOYSA-N
Synonym 2-2-oxocyclopentyl acetic acid, 2-oxocyclopentaneacetic acid, 2-oxo-cyclopentaneacetic acid, 2-oxocyclopentylacetic acid, 2-oxocyclopentyl acetic acid, 2-oxo-cyclopentaneaceticacid, acmc-1bx7z, 2-oxo-cyclopentyl acetic acid, cyclopentaneaceticacid, 2-oxo, 2-2-oxocyclopentyl aceticacid
PubChem CID 3569983
IUPAC Name 2-(2-oxocyclopentyl)acetic acid
SMILES C1CC(C(=O)C1)CC(=O)O

Specifications

Melting Point 47°C to 51°C
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material 2-Oxocyclopentylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.