missing translation for 'onlineSavingsMsg'
Learn More

2-Oxobutanoic acid, MedChemExpress

Product Code. 16483419
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483419 Supplier Medchem Express Supplier No. HYW007926
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Oxobutanoic acid is a product in the enzymatic cleavage of cystathionine.

2-Oxobutanoic acid is a product in the enzymatic cleavage of cystathionine.

Specifications

Chemical Name or Material 2-Oxobutanoic acid
CAS 600-18-0
Color Colorless
Purity Grade Notes Research
Molecular Formula C4H6O3
Quantity 1 g
Solubility Information DMSO : ≥ 50 mg/mL (489.76 mM)
SMILES CCC(=O)C(O)=O
Molecular Weight (g/mol) 102.09
Formula Weight 102.09
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.