missing translation for 'onlineSavingsMsg'
Learn More

2-Oxo-2,3-dihydrobenzo[d]oxazol-6-ylboronic Acid Pinacol Ester, TRC

Product Code. 30424905
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424905 5 mg -
30409995 10 mg 10mg
30409876 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424905 Supplier Toronto Research Chemicals Supplier No. TRCO9903185MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1105710-32-4
Molecular Formula C13H16BNO4
Molecular Weight (g/mol) 261.08
Synonym 2-Oxo-2,3-dihydrobenzoxazole-6-boronic Acid Pinacol Ester
IUPAC Name 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-1,3-benzoxazol-2-one
SMILES CC1(C)OB(OC1(C)C)c2ccc3NC(=O)Oc3c2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H16BNO4/c1-12(2)13(3,4)19-14(18-12)8-5-6-9-10(7-8)17-11(16)15-9/h5-7H,1-4H3,(H,15,16)
Formula Weight 261.1172
Chemical Name or Material 2-Oxo-2,3-dihydrobenzo[d]oxazol-6-ylboronic Acid Pinacol Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.