missing translation for 'onlineSavingsMsg'
Learn More

2-Oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carbaldehyde, TRC

Product Code. 30423204
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423204 500 mg 500mg
30423203 100 mg -
30418090 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423204 Supplier Toronto Research Chemicals Supplier No. TRCO990210500MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106429-59-8
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.14
Synonym 2,3-Dihydro-2-oxo-1H-benzimidazole-5-carboxaldehyde
IUPAC Name 2-hydroxy-3H-benzimidazole-5-carbaldehyde
SMILES Oc1nc2ccc(C=O)cc2[nH]1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H6N2O2/c11-4-5-1-2-6-7(3-5)10-8(12)9-6/h1-4H,(H2,9,10,12)
Formula Weight 162.0429
Chemical Name or Material 2-Oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.