missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%

Product Code. 15450339
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450339 1 g 1g
15470339 250 mg 250mg
15460339 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450339 Supplier Thermo Scientific Alfa Aesar Supplier No. H63578.03
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1159599-99-1 | C7H11NO5 | 189.17 g/mol

2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1159599-99-1
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.17
MDL Number MFCD11976178
InChI Key KOUVDKDABFOPIG-UHFFFAOYSA-N
Synonym 2-oxa-6-azaspiro 3.3 heptane oxalate, 2-oxa-6-aza-spiro 3.3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3,3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3.3 heptane xoxalate, 2-oxa-6-azaspiro 3.3 heptane oxalic acid, 2-oxa-6-azaspiro 3.3 heptane; oxalic acid, 2-oxa-6-azaspiro 3.3 heptane ethanedioate, 2-oxa-6-azaspiro 3.3 heptane oxalate 1:1, 2-oxa-6-azaspiro 3.3 heptane, ethanedioate 1:1, acmc-2099rj
PubChem CID 53308464
SMILES OC(=O)C(O)=O.C1NCC11COC1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
IUPAC Name 2-oxa-6-azaspiro[3.3]heptane;oxalic acid
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 2-Oxa-6-azaspiro[3.3]heptane oxalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.