Learn More
Thermo Scientific™ 2'-OMe-Bz-A Phosphoramidite, standard grade, 20-400 finish bottle
Shop All Thermo Scientific ProductsDescription
2'-OMe-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.
Features:
• HPLC purity—≥98.0%; Sum of non-primary peaks 140-152ppm: ≤0.5%
Specifications
Specifications
| 31P NMR Purity | ≥98.0% |
| Format | Amber bottle (20-400 finish neck style) |
| For Use With (Equipment) | Expedite Synthesizers |
| Description | Base: A Base Protection: Bz |
| Molecular Weight (g/mol) | 887.97 g/mol |
| Product Type | 2in-OMe-Bz-A CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Adenosine (A) |
| Reagent Features | 2in-OMe modified |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.