missing translation for 'onlineSavingsMsg'
Learn More

2-Octynoic acid, 98%

Product Code. 11435067
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435067 100 g 100g
11415067 5 g 5g
11425067 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435067 Supplier Thermo Scientific Alfa Aesar Supplier No. B21957.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5663-96-7 | C8H12O2 | 140.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5663-96-7
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00004364
InChI Key BQDKCWCMDBMLEH-UHFFFAOYSA-N
Synonym 2-octynoic acid, 2-octyn-1-oic acid, m-pentyl propiolate, propiolic acid, pentyl, propiolic acid, pentyl ester, pentylpropiolic acid, 2-oa, 2-octynoicacid, acmc-1apg2, epitope id:137475
PubChem CID 21872
ChEBI CHEBI:60295
IUPAC Name oct-2-ynoic acid
SMILES CCCCCC#CC(=O)O

Specifications

Density 0.961
Boiling Point 148°C to 149°C (19 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.46
Quantity 100 g
UN Number UN3265
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material 2-Octynoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.