missing translation for 'onlineSavingsMsg'
Learn More

2-Octyne, 97%, stabilized, Thermo Scientific™

Product Code. 15395018 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15395018 5 g 5g
15305028 25 g 25g
15315028 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15395018 Supplier Thermo Scientific Acros Supplier No. 455570050
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2809-67-8
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00027302
InChI Key QCQALVMFTWRCFI-UHFFFAOYSA-N
Synonym 2-octyne, n-c5h11c.$.cch3, amylmethylacetylene, methylpentylacetylene, acmc-209h0u, n-c5h11c.equiv.cch3
PubChem CID 17769
IUPAC Name oct-2-yne
SMILES CCCCCC#CC

Specifications

Density 0.7590g/mL
Boiling Point 137.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 0.759
Stabilizer Approx. 200ppm
Formula Weight 110.2
Percent Purity 97%
Chemical Name or Material 2-Octyne
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Octyne
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.