missing translation for 'onlineSavingsMsg'
Learn More

2-Octen-4-one, 97%

Product Code. 11405906
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11405906

Brand: Thermo Scientific Alfa Aesar B22250.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4643-27-0 | C8H14O | 126.199 g/mol

Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4643-27-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00061023
InChI Key FMDLEUPBHMCPQV-GQCTYLIASA-N
Synonym 2-octen-4-one, propenyl butyl ketone, butyl propenyl ketone, unii-c9nb51lmxt, 2-octen-4-one natural, fema no. 3603, c9nb51lmxt, e-oct-2-en-4-one, 2e-oct-2-en-4-one, 2e-2-octen-4-one
PubChem CID 5365891
IUPAC Name (E)-oct-2-en-4-one
SMILES CCCCC(=O)C=CC

Specifications

Density 0.85
Boiling Point 65°C (15 mmHg)
Flash Point 58°C (136°F)
Refractive Index 1.445
Quantity 1 g
UN Number UN1224
Solubility Information Insoluble in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 2-Octen-4-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.