missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone, 98%

Product Code. 11444753
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
Unit Size:
1000mL
250mL
5000mL
This item is not returnable. View return policy

Product Code. 11444753

Brand: Thermo Scientific Alfa Aesar A11643.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-13-7 | C8H16O | 128.215 g/mol

2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-13-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00009540
InChI Key ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
Synonym 2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af
PubChem CID 8093
ChEBI CHEBI:87434
IUPAC Name octan-2-one
SMILES CCCCCCC(=O)C

Specifications

Melting Point -16°C
Density 0.819
Boiling Point 172°C to 173°C
Flash Point 54°C (129°F)
Refractive Index 1.415
Quantity 250 mL
Beilstein 635843
Merck Index 14,4711
Solubility Information Slightly soluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 2-Octanone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.