missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone, 98%

Product Code. 11434753
Change view
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
Unit Size:
1000mL
250mL
5000mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434753 1000 mL 1000mL
11444753 250 mL 250mL
11454753 5000 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434753 Supplier Thermo Scientific Alfa Aesar Supplier No. A11643.AU
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-13-7 | C8H16O | 128.215 g/mol

2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-13-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00009540
InChI Key ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
Synonym 2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af
PubChem CID 8093
ChEBI CHEBI:87434
IUPAC Name octan-2-one
SMILES CCCCCCC(=O)C

Specifications

Melting Point -16°C
Boiling Point 172°C to 173°C
Flash Point 63°C (145°F)
Quantity 1000 mL
Beilstein 635843
Merck Index 14,4711
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 2-Octanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.