missing translation for 'onlineSavingsMsg'
Få mere at vide

2'-O-Methylguanosine, 99%, Thermo Scientific Chemicals

Artikelnummer. 15467069
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 g
Pakningsstørrelse:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
15467069 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15467069 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. J65770.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2140-71-8
Molecular Formula C11H15N5O5
Molecular Weight (g/mol) 297.27
MDL Number MFCD00057053
InChI Key OVYNGSFVYRPRCG-YPLCUDRINA-N
Synonym 2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine
PubChem CID 188959
ChEBI CHEBI:19229
SMILES CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O

Tekniske data

Color White
Quantity 1 g
IUPAC Name 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-3H-purin-6-one
Formula Weight 297.27
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-O-Methylguanosine
compliance-icons
Produktidentifikator
  • 2'-O-Methylguanosine
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplerende oplysninger
  • MIXTURE LIST-Contain: 2'-O-Methylguanosine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.