missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-O-Methylcytidine, 99%

Product Code. 15406959
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15406959

Brand: Thermo Scientific Chemicals J65528.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2140-72-9 | C10H15N3O5 | 257.246 g/mol

2'-O-Methylcytidine is used in the preparation of nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-O-Methylcytidine is used in the preparation of nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

Solubility
Soluble in DMSO.

Notes
Light & Air Sensitive
TRUSTED_SUSTAINABILITY

Specifications

CAS 2140-72-9
Color White
Molecular Formula C10H15N3O5
MDL Number MFCD00056067
Quantity 1 g
Synonym 2'-o-methylcytidine, cytidine, 2'-o-methyl, 2'-o-methyl cytidine, o 2'-methylcytidine, 4-amino-1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidin-2 1h-one, 2'-o-methyl-cytidine, o-2-methylcytidine, o2'-methylcytidine, 2-o-methylcytidine, 2/'-o-methylcytidine
Solubility Information Soluble in DMSO.
InChI Key RFCQJGFZUQFYRF-ZOQUXTDFSA-N
SMILES COC1C(C(OC1N2C=CC(=NC2=O)N)CO)O
IUPAC Name 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidin-2-one
Molecular Weight (g/mol) 257.246
PubChem CID 150971
ChEBI CHEBI:19228
Formula Weight 257.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-O-Methylcytidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.