missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-O-Methyladenosine, 99%

Product Code. 15446649
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446649 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446649 Supplier Thermo Scientific Chemicals Supplier No. J64922.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2140-79-6 | C11H15N5O4 | 281.27 g/mol

2'-O-Methyladenosine is an analog of adenosine used to prepare nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-O-Methyladenosine is an analog of adenosine used to prepare nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

Solubility
Soluble in water.

Notes
Store away from st
TRUSTED_SUSTAINABILITY

Specifications

CAS 2140-79-6
Color White
Molecular Formula C11H15N5O4
MDL Number MFCD00056002
Quantity 1 g
Synonym 2'-o-methyladenosine, adenosine, 2'-o-methyl, 2'-o-methyl adenosine, 2r,3r,4r,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl-4-methoxytetrahydrofuran-3-ol, unii-02yx82ihz5, chembl73237, 2'-o-methyl-adenosine, 2r,3r,4r,5r-5-6-aminopurin-9-yl-2-hydroxymethyl-4-methoxyoxolan-3-ol, 2r,3r,4r,5r-5-6-aminopurin-9-yl-2-hydroxymethyl-4-methoxy-tetrahydrofuran-3-ol, cordysinin b
Solubility Information Soluble in water.
InChI Key FPUGCISOLXNPPC-IOSLPCCCSA-N
SMILES CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=CN=C12
IUPAC Name (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
Molecular Weight (g/mol) 281.27
PubChem CID 102213
ChEBI CHEBI:69426
Formula Weight 281.27
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-O-Methyladenosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.