missing translation for 'onlineSavingsMsg'
Learn More

2-(o-Hydroxyphenyl)benzoxazole,MP Biomedicals

Product Code. 11461306 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461306 10 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11461306 Supplier MP Biomedicals Supplier No. 0520471505
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 835-64-3
Molecular Formula C13H9NO2
Quantity 10 g
Synonym 2-2-hydroxyphenyl benzoxazole, 6-3h-1,3-benzoxazol-2-ylidene cyclohexa-2,4-dien-1-one, 2-2-benzoxazolyl phenol, ghgzvwotjdlrey-uhfffaoysa-n, phenol, benzoxazolyl, acmc-209psp, 2-2'-hydroxyphenyl benzoxazole, 2,4-cyclohexadien-1-one, 6-2 3h-benzoxazolylidene
InChI Key UWUSYRFTCSSHAJ-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2
IUPAC Name 6-(3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol) 211.22
PubChem CID 6744797
Formula Weight 211.22
Chemical Name or Material 2-(O-Hydroxyphenyl)Benzoxazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.