missing translation for 'onlineSavingsMsg'
Learn More

2-Nonanone, 97%, Thermo Scientific Chemicals

Product Code. 10553501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10553501 100 mL Glass bottle 100mL
10123340 5 mL Glass bottle 5mL
10227403 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10553501 Supplier Thermo Scientific Acros Supplier No. 148781000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 821-55-6 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-55-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009553
InChI Key VKCYHJWLYTUGCC-UHFFFAOYSA-N
Synonym 2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
PubChem CID 13187
ChEBI CHEBI:77927
IUPAC Name nonan-2-one
SMILES CCCCCCCC(=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -21.0°C
Color Yellow
Density 0.8260g/mL
Boiling Point 188.0°C to 190.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)6COCH3
Refractive Index 1.4200 to 1.4220
Quantity 100 mL
Beilstein 01, 709
Specific Gravity 0.826
Solubility Information Solubility in water: 0.37g/L (25°C). Other solubilities: soluble in acetone
Formula Weight 142.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Nonanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.