missing translation for 'onlineSavingsMsg'
Learn More

2-Nonanone, 98+%

Product Code. 11309569
Click to view available options
Quantity:
50 g
100 g
1000 g
Unit Size:
1000g
100g
50g
This item is not returnable. View return policy

Product Code. 11309569

Brand: Thermo Scientific Alfa Aesar A11781.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 821-55-6 | C9H18O | 142.242 g/mol

2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-55-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00009553
InChI Key VKCYHJWLYTUGCC-UHFFFAOYSA-N
Synonym 2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
PubChem CID 13187
ChEBI CHEBI:77927
IUPAC Name nonan-2-one
SMILES CCCCCCCC(=O)C

Specifications

Melting Point -15°C
Density 0.826
Boiling Point 195°C
Flash Point 64°C (147°F)
Refractive Index 1.421
Quantity 100 g
Beilstein 1743645
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 142.24
Percent Purity ≥98%
Chemical Name or Material 2-Nonanone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.