missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrothiophene, tech. 85%

Product Code. 11478057
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478057 5 g 5g
11488057 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478057 Supplier Thermo Scientific Alfa Aesar Supplier No. A17464.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 609-40-5 | C4H3NO2S | 129.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-40-5
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.133
MDL Number MFCD00005425
InChI Key JIZRGGUCOQKGQD-UHFFFAOYSA-N
Synonym thiophene, 2-nitro, 2-nitro-thiophene, 2-nitro thiophene, 2-nitrothiophene, tech, nitrothiophene, 2-nitrothiofene, pubchem5570, 2-nitrothiophene, tech., acmc-1aw2t, wln: t5sj bnw
PubChem CID 11866
IUPAC Name 2-nitrothiophene
SMILES C1=CSC(=C1)[N+](=O)[O-]

Specifications

Melting Point 43°C to 45°C
Density 1.3644
Boiling Point 224°C to 225°C
Flash Point 94°C (201°F)
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 129.14
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Nitrothiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.