missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrothiophene-4-carboxylic acid, 98%, Thermo Scientific™

Product Code. 11453474
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453474 1 g 1g
11329569 250 mg 250mg
11463474 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453474 Supplier Thermo Scientific Alfa Aesar Supplier No. A10705.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40357-96-8
Molecular Formula C5H3NO4S
Molecular Weight (g/mol) 173.142
MDL Number MFCD00052582
InChI Key VNJUNWUOAKEIKG-UHFFFAOYSA-N
Synonym 5-nitrothiophene-3-carboxylic acid, 2-nitrothiophene-4-carboxylic acid, 5-nitro-thiophene-3-carboxylic acid, 5-nitro-3-thiophenecarboxylic acid, 3-thiophenecarboxylic acid, 5-nitro, maybridge1_003249, acmc-1ascr, 2-nitro-4-carboxythiophene, ksc238c5f, 2-nitro-4-thiophenecarboxylic acid
PubChem CID 2777267
SMILES C1=C(SC=C1C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 145°C to 148°C
Quantity 1 g
Formula Weight 173.15
Percent Purity 98%
Chemical Name or Material 2-Nitrothiophene-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.