missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroso Riluzole, TRC

Product Code. 30389322
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389322 10 mg 10mg
30396861 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389322 Supplier Toronto Research Chemicals Supplier No. TRCH95333110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Nitroso Riluzole
Analyte or Component Names 2-Nitroso Riluzole
Shipping Condition Room Temperature
Molecular Formula C8 H3 F3 N2 O2 S
Synonym 2-Nitroso-6-(trifluoromethoxy)benzo[d]thiazole
InChI Formula InChI=1S/C8H3F3N2O2S/c9-8(10,11)15-4-1-2-5-6(3-4)16-7(12-5)13-14/h1-3H
Solution Type Neat
SMILES FC(F)(F)Oc1ccc2nc(N=O)sc2c1
IUPAC Name 2-nitroso-6-(trifluoromethoxy)-1,3-benzothiazole
Industry Type Pharmaceutical
Molecular Weight (g/mol) 248.182
Quantity 10 mg
Formula Weight 247.987
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.