missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylselenocyanate, 98%

Product Code. 10626671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10626671 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10626671 Supplier Thermo Scientific Acros Supplier No. 293580010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51694-22-5 | C7H4N2O2Se | 227.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51694-22-5
CAS Min % 98.5
CAS Max % 100.0
Melting Point 138°C to 142°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H4N2O2Se
Linear Formula SeCNC6H4NO2
MDL Number MFCD00043146
Quantity 1 g
Synonym selenocyanic acid, 2-nitrophenyl ester, 2-nitrophenyl selenocyanate, 2-nitrophenylselenocyanate, o-nitrophenyl selenocyanate, 1-nitro-2-selenocyanatobenzene, 2-nitrophenyl selanyl formonitrile, 2-nitrophenyl selenocyanic acid, ortho-nitrophenyl selenocyanate, acmc-20anow, o-nitrophenylselenocyanate
InChI Key LHBLJWULWKQRON-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])[Se]C#N
IUPAC Name (2-nitrophenyl) selenocyanate
Molecular Weight (g/mol) 227.08
PubChem CID 103931
Formula Weight 227.08
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenylselenocyanate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Nitrophenylselenocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.