missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylselenocyanate, 97%

Product Code. 11349026
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349026 5 g 5g
11339026 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349026 Supplier Thermo Scientific Alfa Aesar Supplier No. L20193.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51694-22-5 | C7H4N2O2Se | 227.092 g/mol

2-Nitrophenylselenocyanate is used to study the mechanism of its reaction with the zinc/thiolate clusters of metallothionein. It is also used in the preparation of 2,3-seco-5 alfa-cholestane-2,3-diol and in the preparation of 4alfa-methyl-2,3-seco-5 alfa-cholestane-2,3-diol. Further, it is used in the preparation of 2-nitrophenylselenyl derivative and 3alfa-[(2-Nitrophenyl)seleno]androsta-1,5-dien-17beta-ol. In addition to this, it reacts with 6-bromo-hexanoic acid methyl ester to get 6-(2-nitro-phenylselanyl)-hexanoic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrophenylselenocyanate is used to study the mechanism of its reaction with the zinc/thiolate clusters of metallothionein. It is also used in the preparation of 2,3-seco-5 alfa-cholestane-2,3-diol and in the preparation of 4alfa-methyl-2,3-seco-5 alfa-cholestane-2,3-diol. Further, it is used in the preparation of 2-nitrophenylselenyl derivative and 3alfa-[(2-Nitrophenyl)seleno]androsta-1,5-dien-17beta-ol. In addition to this, it reacts with 6-bromo-hexanoic acid methyl ester to get 6-(2-nitro-phenylselanyl)-hexanoic acid methyl ester.

Solubility
Soluble in dichloromethane, tetrahydrofuran, dimethyl formamide and ethanol.

Notes
Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51694-22-5
Molecular Formula C7H4N2O2Se
Molecular Weight (g/mol) 227.092
MDL Number MFCD00043146
InChI Key LHBLJWULWKQRON-UHFFFAOYSA-N
Synonym selenocyanic acid, 2-nitrophenyl ester, 2-nitrophenyl selenocyanate, 2-nitrophenylselenocyanate, o-nitrophenyl selenocyanate, 1-nitro-2-selenocyanatobenzene, 2-nitrophenyl selanyl formonitrile, 2-nitrophenyl selenocyanic acid, ortho-nitrophenyl selenocyanate, acmc-20anow, o-nitrophenylselenocyanate
PubChem CID 103931
IUPAC Name (2-nitrophenyl) selenocyanate
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])[Se]C#N

Specifications

Melting Point 134°C to 141°C
Quantity 5 g
UN Number UN3283
Beilstein 1309777
Solubility Information Soluble in dichloromethane,tetrahydrofuran,dimethyl formamide and ethanol.
Formula Weight 227.08
Percent Purity 97%
Chemical Name or Material 2-Nitrophenylselenocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.