missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl isocyanate, 99%

Product Code. 10319310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10319310 5 g Glass bottle 5g
10522471 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10319310 Supplier Thermo Scientific Supplier No. 296690050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3320-86-3 | C7H4N2O3 | 164.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3320-86-3
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
InChI Key JRVZITODZAQRQM-UHFFFAOYSA-N
Synonym 2-nitrophenyl isocyanate, o-nitrophenyl isocyanate, benzene, 1-isocyanato-2-nitro, isocyanic acid, o-nitrophenyl ester, 2-nitrophenylisocyanate, 2-nitrobenzenisocyanate, nitrophenyl isocyanate, 2-nitrophenylisocynate, 2-nitro phenyl isocyanate, acmc-1cp5k
PubChem CID 76834
IUPAC Name 1-isocyanato-2-nitrobenzene
SMILES C1=CC=C(C(=C1)N=C=O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 38.0°C to 41.0°C
Color Yellow
Boiling Point 135.0°C to 137.0°C (17.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4NCO
Quantity 5 g
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene
Formula Weight 164.12
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2-Nitrophenyl isocyanate
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 2-Nitrophenyl isocyanate
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.