missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl-beta-D-galactopyranoside, 99%

Product Code. 10225370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10225370 5 g Glass bottle 5g
10513681 1 g Glass bottle 1g
10733334 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10225370 Supplier Thermo Scientific Acros Supplier No. 128820050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 369-07-3 | C12H15NO8 | 301.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-07-3
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00063255
InChI Key KUWPCJHYPSUOFW-YBXAARCKSA-N
Synonym 2-nitrophenyl-beta-d-galactopyranoside, onpg, 2-nitrophenyl beta-d-galactopyranoside, 2-nitrophenyl beta-d-galactoside, o-nitrophenyl beta-d-galactopyranoside, o-nitrophenyl-beta-d-galactoside, 1-o-o-nitrophenyl-beta-d-galactopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy tetrahydro-2h-pyran-3,4,5-triol, 2-nitrophenyl b-d-galactopyranoside
PubChem CID 96647
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(2-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 195.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition −70° (20°C c=1,H2O)
Specific Rotation − 70.00
Formula Weight 301.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Nitrophenyl-β-D-galactopyranoside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.