missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl-beta-D-galactopyranoside, 98+%

Product Code. p-7045494
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11323086

Brand: Thermo Scientific Alfa Aesar L10939.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 369-07-3 | C12H15NO8 | 301.251 g/mol

2-Nitrophenyl-beta-D-galactopyranoside is a β-Galactosidase substrate for colorimetric and EIA applications; counterpart of widely employed pNPP/alkaline phosphatase substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrophenyl-beta-D-galactopyranoside is a β-Galactosidase substrate for colorimetric and EIA applications; counterpart of widely employed pNPP/alkaline phosphatase substrate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 20°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-07-3
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00063255
InChI Key KUWPCJHYPSUOFW-YBXAARCKSA-N
Synonym 2-nitrophenyl-beta-d-galactopyranoside, onpg, 2-nitrophenyl beta-d-galactopyranoside, 2-nitrophenyl beta-d-galactoside, o-nitrophenyl beta-d-galactopyranoside, o-nitrophenyl-beta-d-galactoside, 1-o-o-nitrophenyl-beta-d-galactopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy tetrahydro-2h-pyran-3,4,5-triol, 2-nitrophenyl b-d-galactopyranoside
PubChem CID 96647
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(2-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point ∽192°C (decomposition)
Quantity 1 g
Beilstein 92207
Solubility Information Soluble in water.
Optical Rotation −68° (c=1 in Water)
Formula Weight 301.25
Percent Purity ≥98%
Chemical Name or Material 2-Nitrophenyl-beta-D-galactopyranoside

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.