missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenol, 99%

Product Code. 10376720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376720 500 g Glass bottle 500g
10533871 5 g Glass bottle 5g
10316680 100 g Glass bottle 100g
10705411 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10376720 Supplier Thermo Scientific Acros Supplier No. 128745000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-75-5 | C6H5NO3 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-75-5
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00011688
InChI Key IQUPABOKLQSFBK-UHFFFAOYSA-N
Synonym o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
PubChem CID 6947
ChEBI CHEBI:16260
IUPAC Name 2-nitrophenol
SMILES OC1=CC=CC=C1[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 43.0°C to 47.0°C
Color Yellow
Boiling Point 214.0°C to 216.0°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4OH
Quantity 500 g
UN Number 1663
Beilstein 06, 213
Fieser 05,477
Merck Index 15, 6707
Solubility Information Solubility in water: 2g/L (25°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene,carbon disulfide,alkali hydroxides,and toluene
Viscosity 1.35 mPa.s (80°C)
Formula Weight 139.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Nitrophenol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.