missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroimidazole, 98%

Product Code. 10095903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095903 1 g Glass bottle 1g
10645072 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10095903 Supplier Thermo Scientific Acros Supplier No. 187570010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 527-73-1 | C3H3N3O2 | 113.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-73-1
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00005185
InChI Key YZEUHQHUFTYLPH-UHFFFAOYSA-N
Synonym 2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole
PubChem CID 10701
ChEBI CHEBI:67135
IUPAC Name 2-nitro-1H-imidazole
SMILES [O-][N+](=O)C1=NC=CN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 287°C
Color Green-Khaki to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 914
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alkaline h2o- insoluble in acidic h2o,soluble in methanol,ethanol,acetone,ethyl,acetate and butyl acetate,practically insoluble in ether and chloroform
Formula Weight 113.08
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Nitroimidazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitroimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.