missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrodibenzothiophene, TRC

Product Code. 30483353
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30483353 10 g 10g
30481671 1 g -
2 options
This item is not returnable. View return policy

Product Code. 30483353

Brand: Toronto Research Chemicals TRCN49583510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6639-36-7
Formula Weight 229.0197
InChI Formula InChI=1S/C12H7NO2S/c14-13(15)8-5-6-12-10(7-8)9-3-1-2-4-11(9)16-12/h1-7H
IUPAC Name 2-nitrodibenzothiophene
Molecular Formula C12 H7 N O2 S
Molecular Weight (g/mol) 229.25
SMILES [O-][N+](=O)c1ccc2sc3ccccc3c2c1
Synonym Dibenzothiophene, 2-nitro- (6CI, 7CI, 8CI, 9CI, ACI), 2-Nitrodibenzothiophene (ACI), 2-Nitrodibenzo[b,d]thiophene, NSC 16062
Chemical Name or Material 2-Nitrodibenzothiophene

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.