missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrocinnamaldehyde, predominantly trans, 98%

Product Code. 11484053
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484053 10 g 10g
11359569 25 g 25g
11494053 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484053 Supplier Thermo Scientific Alfa Aesar Supplier No. A10308.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1466-88-2 | C9H7NO3 | 177.159 g/mol

It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1466-88-2
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00007188
InChI Key VMSMELHEXDVEDE-HWKANZROSA-N
Synonym 2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
PubChem CID 5367122
IUPAC Name (E)-3-(2-nitrophenyl)prop-2-enal
SMILES C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]

Specifications

Melting Point 125°C to 129°C
Odor Characteristic
Quantity 10 g
Beilstein 908376
Sensitivity Air Sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 2-Nitrocinnamaldehyde, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.