missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrocinnamaldehyde, predominantly trans, 98%, Thermo Scientific Chemicals

Catalog Number p-7080674
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy
1466-88-2
C9H7NO3
177.159
MFCD00007188
VMSMELHEXDVEDE-HWKANZROSA-N
2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
5367122
(E)-3-(2-nitrophenyl)prop-2-enal
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
This item is not returnable. View return policy
1466-88-2
C9H7NO3
177.159
MFCD00007188
VMSMELHEXDVEDE-HWKANZROSA-N
2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
5367122
(E)-3-(2-nitrophenyl)prop-2-enal
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]

It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 125°C to 129°C
Odor Characteristic
Quantity 10 g
Beilstein 908376
Sensitivity Air Sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 2-Nitrocinnamaldehyde, predominantly trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'