missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrochalcone, 97%

Product Code. 11493358
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493358 25 g 25g
11483358 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493358 Supplier Thermo Scientific Alfa Aesar Supplier No. A17763.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7473-93-0 | C15H11NO3 | 253.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7473-93-0
Molecular Formula C15H11NO3
Molecular Weight (g/mol) 253.257
MDL Number MFCD00031069
InChI Key KTXHLWZQKQDFRF-ZHACJKMWSA-N
Synonym 2-nitrochalcone, chalcone, 2-nitro, 2-nitrobenzylideneacetophenone, 2e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, ccris 1671, e-3-2-nitro-phenyl-1-phenyl-propenone, 2-propen-1-one, 3-2-nitrophenyl-1-phenyl, e-3-2-nitrophenyl-1-phenyl-2-propen-1-one, e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, e-3-2-nitrophenyl-1-phenyl-prop-2-en-1-one
PubChem CID 5337611
IUPAC Name (E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2[N+](=O)[O-]

Specifications

Melting Point 124°C to 128°C
Quantity 25 g
Formula Weight 253.26
Percent Purity 97%
Chemical Name or Material 2-Nitrochalcone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.