missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzylcyanide, TRC

Product Code. 30482269
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30482269 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30482269

Brand: Toronto Research Chemicals TRCN4954251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Nitrobenzylcyanide
CAS 610-66-2
Molecular Formula C8 H6 N2 O2
Quantity 1 g
Synonym Benzeneacetonitrile','2-nitro-, Acetonitrile','(o-nitrophenyl)- (7CI,8CI), 2-Nitrobenzeneacetonitrile, (2-Nitrophenyl)acetonitrile, (o-Nitrophenyl)acetonitrile, 2-Nitrobenzyl cyanide, NSC 99338, o-Nitrobenzacetonitrile, o-Nitrobenzyl cyanide, 2-Nitrophenylacetonitrile
InChI Formula InChI=1S/C8H6N2O2/c9-6-5-7-3-1-2-4-8(7)10(11)12/h1-4H,5H2
SMILES [O-][N+](=O)c1ccccc1CC#N
IUPAC Name 2-(2-nitrophenyl)acetonitrile
Molecular Weight (g/mol) 162.15
Formula Weight 162.0429
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.