missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzylamine hydrochloride, 98%

Product Code. 11443148
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443148 5 g 5g
11433148 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443148 Supplier Thermo Scientific Alfa Aesar Supplier No. A18900.06
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24835-08-3 | C7H9ClN2O2 | 188.611 g/mol

2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies. Also reactant involved in synthesis of tetrahydroisoquinolines via Heck-aza-Michael reactions, aminomethylphenylamine scaffolds for factor Xa inhibition, squaryliamides for mannosyltransferase inhibition and fluorescent two-photon absorption benzothiadiazole dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies. Also reactant involved in synthesis of tetrahydroisoquinolines via Heck-aza-Michael reactions, aminomethylphenylamine scaffolds for factor Xa inhibition, squaryliamides for mannosyltransferase inhibition and fluorescent two-photon absorption benzothiadiazole dyes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24835-08-3
Molecular Formula C7H9ClN2O2
Molecular Weight (g/mol) 188.611
MDL Number MFCD00136280
InChI Key BASJTVIZZDEQBJ-UHFFFAOYSA-N
Synonym 2-nitrobenzylamine hydrochloride, 2-nitrophenyl methanamine hydrochloride, 1-2-nitrophenyl methanamine hydrochloride, 2-nitrobenzylamine hcl, 2-nitrobenzyl amine hcl, acmc-1co7n, o-nitrobenzylamine hydrochloride, 2-nitrobenzyl amine hydrochloride, 2-nitrobenzenemethanamine hydrochloride, 2-nitrophenyl methanaminehydrochloride
PubChem CID 12235442
IUPAC Name (2-nitrophenyl)methanamine;hydrochloride
SMILES C1=CC=C(C(=C1)CN)[N+](=O)[O-].Cl

Specifications

Melting Point 250°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 188.62
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.