missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl alcohol, 97%

Product Code. 11482877
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482877 25 g 25g
11492877 100 g 100g
11402887 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482877 Supplier Thermo Scientific Alfa Aesar Supplier No. A15496.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 612-25-9 | C7H7NO3 | 153.14 g/mol

The o-nitrobenzyl group (o-NB) is utilized frequently in polymer and materials science. It covers the use of based cross-linkers for photodegradable hydrogels, o-NB side chain functionalization in (block) copolymers, o-NB side chain functionalization for thin film patterning, o-NB for self-assembled monolayers, photo cleavable block copolymers, and photo cleavable bioconjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The o-nitrobenzyl group (o-NB) is utilized frequently in polymer and materials science. It covers the use of based cross-linkers for photodegradable hydrogels, o-NB side chain functionalization in (block) copolymers, o-NB side chain functionalization for thin film patterning, o-NB for self-assembled monolayers, photo cleavable block copolymers, and photo cleavable bioconjugates.

Solubility
Soluble in water 5000 mg/L 20 deg Celsius.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-25-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007186
InChI Key BWRBVBFLFQKBPT-UHFFFAOYSA-N
Synonym 2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol
PubChem CID 11923
IUPAC Name (2-nitrophenyl)methanol
SMILES OCC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 70°C to 74°C
Boiling Point 270°C
Quantity 25 g
Beilstein 2046649
Sensitivity Light sensitive
Solubility Information Soluble in water 5000mg/L 20 deg Celsius.
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Nitrobenzyl alcohol
compliance-icons
Product Identifier
  • 2-Nitrobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Nitrobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.