missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzoyl chloride, 95%

Product Code. 10582631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10582631 25 g Glass bottle 25g
10286570 5 g Glass bottle 5g
10445485 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10582631 Supplier Thermo Scientific Acros Supplier No. 232310250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 610-14-0 | C7H4ClNO3 | 185.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-14-0
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.57
MDL Number MFCD00007131
InChI Key BWWHTIHDQBHTHP-UHFFFAOYSA-N
Synonym o-nitrobenzoyl chloride, benzoyl chloride, 2-nitro, 2-nitrobenzoylchloride, benzoyl chloride, o-nitro, unii-86w2g0ja5q, nitrobenzoyl chloride, ccris 3135, 2-nitro-benzoyl chloride, benzoyl chloride, nitro, dsstox_cid_5741
PubChem CID 11875
IUPAC Name 2-nitrobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 17°C to 20°C
Color Brown to Yellow
Density 1.4000g/mL
Boiling Point 148°C to 149°C (9.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula O2NC6H4COCl
Refractive Index 1.5666 to 1.5686
Quantity 25 g
Specific Gravity 1.4
Solubility Information Solubility in water: decomposes
Formula Weight 185.57
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 2-Nitrobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.