missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzotrifluoride, 98%, Thermo Scientific™

Product Code. 15334978 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15334978 1 g 1g
15344978 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15334978 Supplier Thermo Scientific Acros Supplier No. 166520010
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 384-22-5
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD00007145
InChI Key NDZJSUCUYPZXPR-UHFFFAOYSA-N
Synonym 2-nitrobenzotrifluoride, 1-nitro-2-trifluoromethyl benzene, o-nitrobenzotrifluoride, 2-nitro-alpha,alpha,alpha-trifluorotoluene, o-trifluoromethyl nitrobenzene, benzene, 1-nitro-2-trifluoromethyl, unii-ovb3l8givj, ccris 2813, benzene, nitro trifluoromethyl, ovb3l8givj
PubChem CID 9795
IUPAC Name 1-nitro-2-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 31.0°C to 32.0°C
Boiling Point 216.0°C to 217.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula O2NC6H4CF3
Quantity 1 g
Beilstein 05,II,251
Formula Weight 191.11
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzotrifluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.