missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfenyl chloride, 97%

Product Code. 11415667
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415667 50 g 50g
11405667 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415667 Supplier Thermo Scientific Alfa Aesar Supplier No. B23627.18
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7669-54-7 | C6H4ClNO2S | 189.613 g/mol

2-Nitrobenzenesulfenyl chloride is used as a reagent for N-protection as N-2-nitrophenylsulfenyl (Nps) derivatives, in amino acids and peptides and nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzenesulfenyl chloride is used as a reagent for N-protection as N-2-nitrophenylsulfenyl (Nps) derivatives, in amino acids and peptides and nucleosides.

Solubility
Reacts with water.

Notes
Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7669-54-7
Molecular Formula C6H4ClNO2S
Molecular Weight (g/mol) 189.613
MDL Number MFCD00007128
InChI Key NTNKNFHIAFDCSJ-UHFFFAOYSA-N
Synonym 2-nitrobenzenesulfenyl chloride, 2-nitrophenylsulfenyl chloride, benzenesulfenyl chloride, 2-nitro, o-nitrobenzenesulfenyl chloride, o-nitrophenylsulfenyl chloride, 2-nitrobenzenesulfenylchloride, o-nitrophenylsulphenyl chloride, o-nitrobenzenesulphenyl chloride, 2-nitrophenyl thiohypochlorite, 2-nitrophenyl sulfanyl chlorane
PubChem CID 24319
IUPAC Name (2-nitrophenyl) thiohypochlorite
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])SCl

Specifications

Melting Point 72°C to 75°C
Quantity 50 g
UN Number UN3261
Beilstein 473741
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 189.62
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzenesulfenyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.