missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroanisole, TRC

Product Code. 30476064
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30476064 100 g
30481200 1 g
30481513 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476064 Supplier Toronto Research Chemicals Supplier No. TRCN491950100G
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-23-6
Molecular Formula C7 H7 N O3
Molecular Weight (g/mol) 153.14
Synonym Benzene, 1-methoxy-2-nitro-, Anisole, o-nitro- (8CI), 1-Methoxy-2-nitrobenzene, 1-Nitro-2-methoxybenzene, 2-Methoxy-1-nitrobenzene, 2-Methoxynitrobenzene, 2-Nitroanisole, 2-Nitromethoxybenzene, NSC 5506, o-Methoxynitrobenzene, o-Nitroanisole, o-Nitrobenzene methyl ether, o-Nitrophenyl methyl ether
IUPAC Name 1-methoxy-2-nitrobenzene
SMILES COc1ccccc1[N+](=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/C7H7NO3/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3
Formula Weight 153.0426
Chemical Name or Material 2-Nitroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.