missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroaniline, TRC

Product Code. 30478430
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478430 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478430 Supplier Toronto Research Chemicals Supplier No. TRCN49177510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 2-Nitroaniline
CAS 88-74-4
Molecular Formula C6 H6 N2 O2
Quantity 10 g
Synonym Benzenamine','2-nitro-, Aniline','o-nitro- (8CI), 2-Nitrobenzenamine, 1-Amino-2-nitrobenzene, 1-Nitro-2-aminobenzene, 2-Aminonitrobenzene, 2-Nitroaniline, 2-Nitrophenylamine, Azoene Fast Orange GR Base, Azoene Fast Orange GR Salt, Azofix Orange GR Salt, Azogene Fast Orange GR, Brentamine Fast Orange GR Base, Brentamine Fast Orange GR Salt, Devol Orange B, Devol Orange Salt B, Fast Orange Base JR, Fast Orange GR Base, Fast Orange O Base, Fast Orange O Salt, Fast Orange Salt JR, Hiltonil Fast Orange GR Base, Hiltosal Fast Orange GR Salt, Hindasol Orange GR Salt, NSC 9796, Natasol Fast Orange GR Salt, ONA, Orange Base Ciba II, Orange Base Irga II, Orange GRS Salt, Orange Salt Ciba II, Orange Salt Irga II, o-Aminonitrobenzene, o-Nitroaniline, o-Nitrobenzenamine, o-Nitrophenylamine
InChI Formula InChI=1S/C6H6N2O2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H,7H2
SMILES Nc1ccccc1[N+](=O)[O-]
IUPAC Name 2-nitroaniline
Molecular Weight (g/mol) 138.12
Formula Weight 138.0429
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.