missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroaniline, 98%

Product Code. 11418903
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418903 250 g 250g
11408903 1000 g 1000g
11428903 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418903 Supplier Thermo Scientific Alfa Aesar Supplier No. A11055.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-74-4 | C6H6N2O2 | 138.126 g/mol

2-Nitroaniline is the main precursor to phenylenediamines, which are converted to benzimidazoles, a family of heterocycles that are key components in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitroaniline is the main precursor to phenylenediamines, which are converted to benzimidazoles, a family of heterocycles that are key components in pharmaceuticals.

Solubility
Soluble in water (0.6 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-74-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00007687
InChI Key DPJCXCZTLWNFOH-UHFFFAOYSA-N
Synonym o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzene
PubChem CID 6946
IUPAC Name 2-nitroaniline
SMILES C1=CC=C(C(=C1)N)[N+](=O)[O-]

Specifications

Melting Point 70°C to 74°C
Density 1.255
Boiling Point 284°C to 286°C
Flash Point 168°C (334°F)
Quantity 250 g
UN Number UN1661
Beilstein 509275
Merck Index 14,6583
Solubility Information Soluble in water (0.6g/L at 20°C).
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 2-Nitroaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: o-Nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.