missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)phenol, 99%, Thermo Scientific™

Catalog Number p-4323274
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
400-99-7
C7H4F3NO3
207.11
MFCD00009791
XZEDEVRSUANQEM-UHFFFAOYSA-N
2-nitro-4-trifluoromethyl phenol, 4-hydroxy-3-nitrobenzotrifluoride, phenol, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethylphenol, 2-nitro-alpha,alpha,alpha-trifluoro-p-cresol, 4-hydroxy-3-nitrobenzotrifluoroide, p-cresol, 2-nitro-alpha,alpha,alpha-trifluoro, 2-nitro-4-trifluoromethyl-phenol, pubchem4150, 4-06-00-02150 beilstein handbook reference
9816
2-nitro-4-(trifluoromethyl)phenol
OC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F
This item is not returnable. View return policy
400-99-7
C7H4F3NO3
207.11
MFCD00009791
XZEDEVRSUANQEM-UHFFFAOYSA-N
2-nitro-4-trifluoromethyl phenol, 4-hydroxy-3-nitrobenzotrifluoride, phenol, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethylphenol, 2-nitro-alpha,alpha,alpha-trifluoro-p-cresol, 4-hydroxy-3-nitrobenzotrifluoroide, p-cresol, 2-nitro-alpha,alpha,alpha-trifluoro, 2-nitro-4-trifluoromethyl-phenol, pubchem4150, 4-06-00-02150 beilstein handbook reference
9816
2-nitro-4-(trifluoromethyl)phenol
OC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F
CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Orange
Density 1.4700g/mL
Boiling Point 92.0°C to 94.0°C (12.0 Torr)
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula O2NC6H3(CF3)OH
Refractive Index 1.5035
Quantity 1 g
Beilstein 06,IV,215
Specific Gravity 1.47
Formula Weight 207.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitro-4-(trifluoromethyl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'