missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)benzyl bromide, 97%

Product Code. 15445567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445567 5 g 5g
15435567 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445567 Supplier Thermo Scientific Alfa Aesar Supplier No. H32802.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 162333-02-0 | C8H5BrF3NO3

Used as intermediates. Also as laboratory reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediates. Also as laboratory reagents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162333-02-0
Molecular Formula C8H5BrF3NO3
MDL Number MFCD03095413

Specifications

Melting Point 36°C to 39°C
Quantity 5 g
UN Number UN3261
Solubility Information Insoluble in water.
Formula Weight 284.03
Percent Purity 97%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.