missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™

Product Code. 10356814
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356814 5 g Amber Glass Bottle 5g
10495332 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10356814 Supplier Thermo Scientific Maybridge Supplier No. TL00377DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2267-23-4
Molecular Formula C7H5F3N2O3
Molecular Weight (g/mol) 222.12
MDL Number MFCD00042326
InChI Key YCGFVAPIBALHRT-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethoxy aniline, 2-amino-5-trifluoromethoxy nitrobenzene, benzenamine, 2-nitro-4-trifluoromethoxy, 1-amino-2-nitro-4-trifluoromethoxy benzene, 2-nitro-4-trifluoromethoxy phenylamine, 4-amino-3-nitro-alpha,alpha,alpha-trifluoroanisole, pubchem8514, acmc-1cnwp, ksc496g9b, 2-nitro4-trifluoromethoxyaniline
PubChem CID 2775772
IUPAC Name 2-nitro-4-(trifluoromethoxy)aniline
SMILES NC1=CC=C(OC(F)(F)F)C=C1[N+]([O-])=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 222.12
Percent Purity 95%
Chemical Name or Material 2-Nitro-4-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.