missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylamine, TRC

Product Code. 30135328
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
1g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135328 25 g 25g
30136029 2.5 g 2.5g
17957869 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30135328 Supplier Toronto Research Chemicals Supplier No. TRCN37801025G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-59-8
Molecular Formula C10 H9 N
Molecular Weight (g/mol) 143.19
Synonym 2-Naphthylamine (8CI), 2-Aminonaphthalene, C.I. 37270, Fast Scarlet Base B, Naphthalen-2-ylamine, o-Aminonaphthalene, β-Aminonaphthalene, β-Naphthylamine
IUPAC Name naphthalen-2-amine
SMILES Nc1ccc2ccccc2c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H9N/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,11H2
Formula Weight 143.0735
Percent Purity >95%
Chemical Name or Material 2-Aminonaphthalene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.